Assay Details
Target Gene Details
Entrez Gene ID: | 360213 |
Gene Name: | tripartite motif-containing 46 |
Gene Aliases: |
TRIFIC |
Location: |
Chr.3:89234169-89246154 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim46 | NM_001039466.1 | NP_001034555.1 | ||
| NM_183037.2 | 9 | 1856 | NP_898858.1 | |
| NR_121683.1 | 10 | 2024 | ||
| AK132368.1 | ||||
| AK170904.1 | BAE42105.1 | |||
| AY251388.1 | 9 | 1736 | AAP51208.1 |
More Information
Set Membership: |
|

Genomic Map