Assay Details
Target Gene Details
Entrez Gene ID: | 26879 |
Gene Name: | UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 1 |
Gene Aliases: |
B3galt3, b3GT3 |
Location: |
Chr.3:69573919-69598960 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3galnt1 | NM_020026.4 | 3 | 520 | NP_064410.1 |
| AK003837.1 | BAB23028.1 | |||
| AK088407.1 | 3 | 423 | BAC40336.1 | |
| BC003835.1 | 2 | 326 | AAH03835.3 | |
| BY278563.1 |
More Information
Set Membership: |
|

Genomic Map