Assay Details
Target Gene Details
Entrez Gene ID: | 242151 |
Gene Name: | potassium voltage-gated channel, shaker-related subfamily, member 10 |
Gene Aliases: |
Gm1962, Kcna8, Kv1.8 |
Location: |
Chr.3:107183143-107195721 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcna10 | NM_001081140.1 | 2 | 394 | NP_001074609.1 |
| AK157965.1 | 2 | 395 | ||
| BC137828.1 | 2 | 483 | ||
| BC137829.1 | 2 | 483 | ||
| BC158036.1 | 2 | 315 |
More Information
Set Membership: |
|

Genomic Map