Assay Details
Target Gene Details
Entrez Gene ID: | 16491 |
Gene Name: | potassium voltage-gated channel, shaker-related subfamily, member 3 |
Gene Aliases: |
Kca1-3, Kv1.3, Mk-3 |
Location: |
Chr.3:107036162-107038129 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcna3 | NM_008418.2 | 1 | 990 | NP_032444.2 |
| BC132349.1 | 1 | 827 | ||
| BC137667.1 | 1 | 827 |
More Information
Set Membership: |
|

Genomic Map