Assay Details
Target Gene Details
Entrez Gene ID: | 16497 |
Gene Name: | potassium voltage-gated channel, shaker-related subfamily, beta member 1 |
Gene Aliases: |
Akr8a8, Kvbeta1.1, mKv(beta)1 |
Location: |
Chr.3:64949049-65378225 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnab1 | NM_010597.4 | 1 | 568 | NP_034727.3 |
| AF033003.1 | 1 | 922 | AAB87085.1 | |
| AK031362.1 | 1 | 569 | BAC27367.1 | |
| AK035994.1 | ||||
| AK138467.1 | 1 | 557 | BAE23670.1 | |
| BC014701.1 | 1 | 293 | AAH14701.1 | |
| U65591.1 | 1 | 164 | AAB37262.1 | |
| X97281.1 | 1 | 552 | CAA65936.1 |

Genomic Map