Assay Details
Target Gene Details
Entrez Gene ID: | 99890 |
Gene Name: | protein arginine N-methyltransferase 6 |
Gene Aliases: |
AW124876, BB233495, Hrmt1l6 |
Location: |
Chr.3:110246104-110250999 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prmt6 | NM_178891.5 | 1 | 2045 | NP_849222.3 |
| NR_024139.2 | ||||
| AK034732.1 | 1 | 2017 | BAC28811.1 | |
| AK075617.1 | 1 | 1501 | ||
| AK087551.1 | BAC39923.1 | |||
| AK087608.1 | ||||
| AK157132.1 | ||||
| AK172003.1 | 1 | 2037 | BAE42769.1 | |
| AK172105.1 | 1 | 2045 | BAE42828.1 | |
| AK172722.1 | 1 | 2045 | BAE43144.1 | |
| BC022899.1 | 1 | 2029 | AAH22899.1 | |
| CF748122.1 |
More Information
Set Membership: |
|

Genomic Map