Assay Details
Target Gene Details
Entrez Gene ID: | 27223 |
Gene Name: | transformation related protein 53 binding protein 1 |
Gene Aliases: |
53BP1, Tp53bp1, m53BP1, p53BP1 |
Location: |
Chr.2:121194835-121290320 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53bp1 | NM_001290830.1 | 19 | 4109 | NP_001277759.1 |
| NM_013735.4 | 19 | 4109 | NP_038763.3 | |
| AJ414734.1 | 18 | 3952 | CAC94013.1 | |
| AK132323.1 | 1 | 142 | BAE21103.1 | |
| BC035206.1 | 7 | 1321 | AAH35206.1 | |
| BC079906.1 | 19 | 4043 | AAH79906.1 |
More Information
Set Membership: |
|

Genomic Map