Assay Details
Target Gene Details
Entrez Gene ID: | 68728 |
Gene Name: | transformation related protein 53 inducible nuclear protein 2 |
Gene Aliases: |
1110029F20Rik, Tp53inp2 |
Location: |
Chr.2:155381808-155389847 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53inp2 | NM_178111.3 | 3 | 528 | NP_835212.1 |
| AJ297793.1 | 3 | 460 | CAC82594.1 | |
| AK146589.1 | 3 | 533 | BAE27284.1 | |
| AK159265.1 | 1 | 104 | BAE34945.1 | |
| AK170132.1 | 3 | 566 | BAE41585.1 | |
| BC036958.1 | 2 | 335 | ||
| BC043086.1 | 3 | 528 | AAH43086.1 |

Genomic Map