Assay Details
Target Gene Details
Entrez Gene ID: | 71643 |
Gene Name: | zinc finger, GRF-type containing 1 |
Gene Aliases: |
4930422G04Rik, AI448607 |
Location: |
Chr.3:127553436-127618023 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zgrf1 | NM_197997.2 | 17 | 4427 | NP_932114.2 |
| AK046429.1 | 12 | 2152 | BAC32725.1 | |
| AK156938.1 | 2 | 420 | BAE33905.1 |

Genomic Map