Assay Details
Target Gene Details
Entrez Gene ID: | 75015 |
Gene Name: | RIKEN cDNA 4930503B20 gene |
Gene Aliases: |
1700013A07Rik, 1700095G21Rik, 4930483N21Rik, Gm1652, Samd13 |
Location: |
Chr.3:146645189-146686801 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930503B20Rik | NM_029144.3 | 1 | 278 | NP_083420.2 |
| AK007081.1 | 1 | 281 | ||
| AK015617.1 | 1 | 274 | BAB29906.1 | |
| AK015685.1 | 1 | 276 | ||
| BC048658.1 | 1 | 278 | AAH48658.1 |
Target Gene Details
Entrez Gene ID: | 100286843 |
Gene Name: | predicted gene 16325 |
Gene Aliases: |
OTTMUSG00000033652 |
Location: |
Chr.3:146641932-146651715 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16325 | NR_045949.1 | 4 | 2499 | |
| AK139061.1 | 4 | 2500 |

Genomic Map