Assay Details
Target Gene Details
Entrez Gene ID: | 71519 |
Gene Name: | cytochrome P450, family 2, subfamily u, polypeptide 1 |
Gene Aliases: |
8430436A10Rik |
Location: |
Chr.3:131290491-131303227 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2u1 | NM_027816.3 | 1 | 373 | NP_082092.2 |
| AK018458.1 | 1 | 349 | BAB31223.1 | |
| AK041477.1 | 1 | 374 | BAC30954.1 | |
| AK142740.1 | 1 | 313 |
More Information
Set Membership: |
|

Genomic Map