Assay Details
Target Gene Details
Entrez Gene ID: | 751864 |
Gene Name: | predicted gene 9733 |
Gene Aliases: |
- |
Location: |
Chr.3:15295103-15332310 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9733 | NM_001076679.2 | 2 | 205 | NP_001070147.1 |
| DQ055452.1 | 2 | 206 |
More Information
Set Membership: |
|

Genomic Map