Assay Details
Target Gene Details
Entrez Gene ID: | 241732 |
Gene Name: | TSPY-like 3 |
Gene Aliases: |
AW212607, Gm36 |
Location: |
Chr.2:153222370-153225441 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspyl3 | NM_198617.2 | 1 | 2573 | NP_941019.2 |
| AK134522.1 | 1 | 2570 | BAE22169.1 | |
| AK148882.1 | 1 | 2518 | BAE28689.1 | |
| AK167722.1 | 1 | 2554 | BAE39764.1 | |
| BC058340.1 | 1 | 2515 | AAH58340.1 |
More Information
Set Membership: |
|

Genomic Map