Assay Details
Target Gene Details
Entrez Gene ID: | 545527 |
Gene Name: | glutamate rich 6 |
Gene Aliases: |
4932431H17Rik, Fam194a |
Location: |
Chr.3:58616300-58665413 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erich6 | NM_001081262.1 | 14 | 2064 | NP_001074731.1 |
| AK133299.1 | 11 | 2666 |

Genomic Map