Assay Details
Target Gene Details
Entrez Gene ID: | 12038 |
Gene Name: | butyrylcholinesterase |
Gene Aliases: |
C730038G20Rik |
Location: |
Chr.3:73635805-73708437 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bche | NM_009738.3 | 2 | 1302 | NP_033868.3 |
| AK050337.1 | 2 | 1303 | BAC34196.1 | |
| AK052200.1 | 2 | 1131 | ||
| BC099977.1 | 2 | 1129 | AAH99977.1 | |
| M99492.1 | 1 | 1030 | AAA37328.1 |
More Information
Set Membership: |
|

Genomic Map