Assay Details
Target Gene Details
Entrez Gene ID: | 16007 |
Gene Name: | cysteine rich protein 61 |
Gene Aliases: |
AI325051, CCN1, Igfbp10 |
Location: |
Chr.3:145646971-145649985 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyr61 | NM_010516.2 | 5 | 1362 | NP_034646.1 |
| AK156723.1 | 5 | 1357 | BAE33819.1 | |
| AK159452.1 | 5 | 1354 | BAE35095.1 | |
| BC066019.1 | 5 | 1342 | AAH66019.1 | |
| M32490.1 | 5 | 1358 | AAA37512.1 |
More Information
Set Membership: |
|

Genomic Map