Assay Details
Target Gene Details
Entrez Gene ID: | 229791 |
Gene Name: | lipid phosphate phosphatase-related protein type 4 |
Gene Aliases: |
A330086D10, D3Bwg0562e, PRG-1, mKIAA0455 |
Location: |
Chr.3:117319145-117360876 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lppr4 | NM_177664.5 | 7 | 2279 | NP_808332.3 |
| AF541279.1 | 7 | 2280 | AAP57768.1 | |
| AK046782.1 | 7 | 2280 | BAC32865.1 | |
| AK079635.1 | 7 | 1613 | BAC37711.1 | |
| AK129149.1 | 7 | 1875 | BAC97959.1 | |
| AY266266.1 | 7 | 2280 | AAP41099.1 | |
| AY266267.1 | 7 | 2280 | AAP41100.1 | |
| BC024711.1 | 1 | 419 | AAH24711.1 | |
| BC137701.1 | 7 | 1874 | ||
| BC137702.1 | 7 | 1875 |
More Information
Set Membership: |
|

Genomic Map