Assay Details
Target Gene Details
Entrez Gene ID: | 73124 |
Gene Name: | golgi integral membrane protein 4 |
Gene Aliases: |
3110027H23Rik, GIMPC, GPP130, Golph4, P138 |
Location: |
Chr.3:75876183-75956949 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golim4 | NM_001291069.1 | 16 | 3154 | NP_001277998.1 |
| NM_175193.6 | 15 | 3070 | NP_780402.1 | |
| AK014086.1 | ||||
| AK033730.1 | 15 | 3058 | ||
| AK050346.1 | 3 | 738 | BAC34202.1 | |
| AK078928.1 | 16 | 3154 | ||
| BC086642.1 | 15 | 2902 | AAH86642.1 | |
| BC089354.1 | 15 | 2952 | AAH89354.1 |

Genomic Map