Assay Details
Target Gene Details
Entrez Gene ID: | 19288 |
Gene Name: | pentraxin related gene |
Gene Aliases: |
AI607804, TSG-14 |
Location: |
Chr.3:66219887-66225806 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptx3 | NM_008987.3 | 2 | 445 | NP_033013.3 |
| AK162281.1 | 1 | 631 | ||
| BC022176.1 | 2 | 445 | AAH22176.1 | |
| BQ442682.1 | 2 | 332 | ||
| CA529293.1 | 2 | 445 | ||
| X83601.1 | 2 | 369 | CAA58580.1 |
Target Gene Details
Entrez Gene ID: | 72789 |
Gene Name: | ventricular zone expressed PH domain-containing 1 |
Gene Aliases: |
2810471M23Rik, Veph |
Location: |
Chr.3:66053558-66297066 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Veph1 | NM_145820.3 | NP_665819.2 | ||
| AB085187.1 | BAC02920.1 | |||
| BC128271.1 |
More Information
Set Membership: |
|

Genomic Map