Assay Details
Target Gene Details
Entrez Gene ID: | 241431 |
Gene Name: | xin actin-binding repeat containing 2 |
Gene Aliases: |
2310003D02Rik, 2310008C07Rik, A530024P18Rik, AI452089, Cmya3, Gm352, Xin2 |
Location: |
Chr.2:67446000-67526620 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xirp2 | NM_001024618.2 | 7 | 6913 | NP_001019789.1 |
| NM_001083919.1 | 7 | 6913 | NP_001077388.1 | |
| DQ011666.1 | 7 | 6914 | AAY44537.1 | |
| EF119712.1 | ||||
| EF119716.1 | 7 | 6913 | ||
| EF122140.1 | 6 | 6668 |
More Information
Set Membership: |
|

Genomic Map