Assay Details
Target Gene Details
Entrez Gene ID: | 213054 |
Gene Name: | GA repeat binding protein, beta 2 |
Gene Aliases: |
1810015F01Rik, 5830427M07, 9430006E19Rik, A430024B14Rik, AV050852, Gabpb2-1 |
Location: |
Chr.3:95181766-95217951 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabpb2 | NM_029885.1 | 1 | 185 | NP_084161.1 |
| NM_172512.2 | 1 | 185 | NP_766100.1 | |
| AK086817.1 | 1 | 129 | BAC39751.1 | |
| AK087979.1 | BAC40072.1 | |||
| AK141382.1 | 1 | 159 | BAE24666.1 | |
| CJ101339.1 | 1 | 185 |
Target Gene Details
Entrez Gene ID: | 100504029 |
Gene Name: | predicted gene, 16740 |
Gene Aliases: |
- |
Location: |
Chr.3:95217457-95226658 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16740 | NR_108026.1 | 1 | 302 | |
| AK020406.1 | 1 | 303 |

Genomic Map