Assay Details
Target Gene Details
Entrez Gene ID: | 19729 |
Gene Name: | solute carrier family 50 (sugar transporter), member 1 |
Gene Aliases: |
MmSWEET1, Rag1ap1, Rga |
Location: |
Chr.3:89268246-89270570 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc50a1 | NM_009057.3 | 6 | 734 | NP_033083.2 |
| AK014305.1 | 6 | 687 | BAB29259.1 | |
| AK045924.1 | 4 | 1047 | ||
| AK079716.1 | 5 | 799 | ||
| AK158862.1 | 5 | 759 | ||
| AK170162.1 | 6 | 685 | BAE41608.1 | |
| AK209820.1 | ||||
| BC014292.1 | 6 | 718 | AAH14292.1 | |
| X96618.1 | 6 | 725 | CAA65438.1 |

Genomic Map