Assay Details
Target Gene Details
Entrez Gene ID: | 320226 |
Gene Name: | coiled-coil domain containing 171 |
Gene Aliases: |
4930418J05Rik, 4930473A06Rik, A330015D16Rik |
Location: |
Chr.4:83525537-83866143 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc171 | NM_001081012.1 | 26 | 4154 | NP_001074481.1 |
| AK039282.1 | 14 | 2850 | BAC30305.1 | |
| BC039810.1 | 8 | 993 | AAH39810.1 |

Genomic Map