Assay Details
Target Gene Details
Entrez Gene ID: | 67735 |
Gene Name: | RIKEN cDNA 4930528A17 gene |
Gene Aliases: |
- |
Location: |
Chr.4:21846327-21848448 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930528A17Rik | NR_028384.1 | 1 | 762 | |
| AK014838.1 | 1 | 764 | ||
| AK015922.1 | 1 | 684 | BAB30033.1 | |
| AK039347.1 | 1 | 746 |
Target Gene Details
Entrez Gene ID: | 66625 |
Gene Name: | PNN interacting serine/arginine-rich |
Gene Aliases: |
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18 |
Location: |
Chr.4:21847583-21876475 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnisr | NM_025669.1 | 1 | 105 | NP_079945.1 |
| BC141031.1 |

Genomic Map