Assay Details
Target Gene Details
Entrez Gene ID: | 50935 |
Gene Name: | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6 |
Gene Aliases: |
ST6GalNAcVI, Siat7f |
Location: |
Chr.2:32599709-32620809 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6galnac6 | NM_001025310.2 | 4 | 528 | NP_001020481.1 |
| NM_001025311.2 | 3 | 438 | NP_001020482.1 | |
| NM_001289547.1 | 5 | 583 | NP_001276476.1 | |
| NM_001289548.1 | 4 | 528 | NP_001276477.1 | |
| NM_001289549.1 | 5 | 712 | NP_001276478.1 | |
| NM_016973.3 | 5 | 583 | NP_058669.1 | |
| NR_110347.1 | 5 | 583 | ||
| AB035123.1 | 5 | 544 | BAA95940.1 | |
| AB035174.1 | 2 | 313 | BAA87036.1 | |
| AK030648.1 | 4 | 496 | BAC27064.1 | |
| AK035447.1 | 3 | 413 | ||
| AK157562.1 | 5 | 686 | ||
| AK159158.1 | 5 | 560 | ||
| BC022924.1 | 5 | 527 | ||
| BY720739.1 | 3 | 438 | ||
| CK617550.1 |

Genomic Map