Assay Details
Target Gene Details
Entrez Gene ID: | 228994 |
Gene Name: | YTH domain family 1 |
Gene Aliases: |
2210410K23Rik, 8030473O16 |
Location: |
Chr.2:180904377-180920936 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ythdf1 | NM_173761.3 | 1 | 50 | NP_776122.1 |
| AK033241.1 | 1 | 59 | ||
| AK046768.1 | 1 | 64 | BAC32861.1 | |
| AK075846.1 | ||||
| AK172582.1 | BAE43080.1 | |||
| BC061479.1 | AAH61479.1 | |||
| BC065050.1 | 1 | 50 | AAH65050.1 |

Genomic Map