Assay Details
Target Gene Details
Entrez Gene ID: | 228994 |
Gene Name: | YTH domain family 1 |
Gene Aliases: |
2210410K23Rik, 8030473O16 |
Location: |
Chr.2:180904377-180920936 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ythdf1 | NM_173761.3 | 5 | 2528 | NP_776122.1 |
| AK046768.1 | 5 | 2542 | BAC32861.1 | |
| AK152264.1 | 2 | 894 | BAE31082.1 | |
| AK168430.1 | 2 | 1137 | BAE40338.1 | |
| BC061479.1 | 5 | 2499 | AAH61479.1 | |
| BC065050.1 | 5 | 2528 | AAH65050.1 |

Genomic Map