Assay Details
Target Gene Details
Entrez Gene ID: | 70762 |
Gene Name: | doublecortin-like kinase 2 |
Gene Aliases: |
6330415M09Rik, AU044875, CL2, CLICK2, Click-II, Dcamkl2 |
Location: |
Chr.3:86786150-86920884 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dclk2 | NM_001195496.1 | 17 | 3672 | NP_001182425.1 |
| NM_001195497.1 | 16 | 3624 | NP_001182426.1 | |
| NM_001195498.1 | 18 | 3688 | NP_001182427.1 | |
| NM_001195499.1 | 18 | 3685 | NP_001182428.1 | |
| NM_027539.5 | 16 | 3627 | NP_081815.3 | |
| AK049179.1 | 16 | 3076 | BAC33590.1 | |
| BC056921.1 | 16 | 3595 | AAH56921.1 | |
| BU700795.1 | 1 | 438 |
More Information
Set Membership: |
|

Genomic Map