Assay Details
Target Gene Details
Entrez Gene ID: | 229877 |
Gene Name: | RAP1, GTP-GDP dissociation stimulator 1 |
Gene Aliases: |
BC011279, GDS1 |
Location: |
Chr.3:138925897-139075201 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rap1gds1 | NM_001040690.2 | 15 | 2465 | NP_001035780.1 |
| NM_001286759.1 | 15 | 2462 | NP_001273688.1 | |
| NM_145544.3 | 14 | 2318 | NP_663519.2 | |
| AK160989.1 | 15 | 2398 | BAE36135.1 | |
| AK166315.1 | 15 | 2398 | BAE38698.1 | |
| AK172052.1 | 15 | 2467 | BAE42801.1 |

Genomic Map