Assay Details
Target Gene Details
Entrez Gene ID: | 74012 |
Gene Name: | RAP2B, member of RAS oncogene family |
Gene Aliases: |
4021402C18Rik, AA408554 |
Location: |
Chr.3:61364507-61368703 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rap2b | NM_028712.4 | 1 | 2100 | NP_082988.1 |
| AK014462.2 | 1 | 2007 | BAB29368.1 | |
| AK045030.1 | 1 | 2014 | BAC32189.1 | |
| AK048795.1 | 1 | 112 | ||
| AK163186.1 | 1 | 1983 | BAE37225.1 |
More Information
Set Membership: |
|

Genomic Map