Assay Details
Target Gene Details
Entrez Gene ID: | 72789 |
Gene Name: | ventricular zone expressed PH domain-containing 1 |
Gene Aliases: |
2810471M23Rik, Veph |
Location: |
Chr.3:66053558-66297066 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Veph1 | NM_145820.3 | 2 | 427 | NP_665819.2 |
| AB085187.1 | 2 | 409 | BAC02920.1 | |
| AK164725.1 | 2 | 339 | ||
| BC128271.1 | 1 | 134 |

Genomic Map