Assay Details
Target Gene Details
Entrez Gene ID: | 14595 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1 |
Gene Aliases: |
B-1,4-GalT, B-1,4-GalT1, GalT, Ggtb, Ggtb2, b4Gal-T1, beta-1,4-GalT, beta-1,4-GalT1, beta4Gal-T1 |
Location: |
Chr.4:40804582-40854537 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt1 | NM_022305.4 | 6 | 4062 | NP_071641.1 |
| AA217591.1 | ||||
| AK154683.1 | 1 | 1205 | ||
| BC053006.1 | 6 | 3523 | AAH53006.1 | |
| D00314.1 | 6 | 3476 | BAA00216.1 | |
| J03880.1 | 6 | 3501 | AAA37297.1 |
More Information
Set Membership: |
|

Genomic Map