Assay Details
Target Gene Details
Entrez Gene ID: | 53418 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2 |
Gene Aliases: |
Ggtb2 |
Location: |
Chr.4:117873000-117883482 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt2 | NM_001253381.1 | 3 | 640 | NP_001240310.1 |
| NM_017377.5 | 3 | 605 | NP_059073.1 | |
| NR_045570.1 | 3 | 605 | ||
| AB019541.2 | 1 | 144 | BAA34385.1 | |
| AF142670.1 | 2 | 249 | AAF22220.1 | |
| AK145250.1 | 3 | 606 | ||
| AK165829.1 | 3 | 599 | BAE38399.1 | |
| BC138567.1 | 2 | 241 | ||
| BC138569.1 | 2 | 241 | ||
| BC145335.1 | 2 | 276 |
More Information
Set Membership: |
|

Genomic Map