Assay Details
Target Gene Details
Entrez Gene ID: | 230991 |
Gene Name: | RIKEN cDNA B930041F14 gene |
Gene Aliases: |
MNCb-3966 |
Location: |
Chr.4:155694342-155696485 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B930041F14Rik | NM_178699.4 | 1 | 1471 | NP_848814.3 |
| AB041660.1 | 1 | 992 | BAA95105.1 | |
| AK046537.1 | 1 | 1473 | BAC32775.1 | |
| AK047240.1 | 1 | 1342 | BAC33003.1 | |
| AK048841.1 | 1 | 1316 | BAC33472.1 |

Genomic Map