Assay Details
Target Gene Details
Entrez Gene ID: | 74711 |
Gene Name: | tubulin tyrosine ligase-like family, member 9 |
Gene Aliases: |
1700016F23Rik, 4930509O20Rik, AW046572 |
Location: |
Chr.2:152950811-153008482 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll9 | NM_001083618.1 | 15 | 1630 | NP_001077087.1 |
| AK006027.1 | 15 | 1626 | ||
| AK015388.1 | 15 | 1609 | ||
| AK041200.1 | 2 | 456 | ||
| AK041612.1 | 2 | 405 |
More Information
Set Membership: |
|

Genomic Map