Assay Details
Target Gene Details
Entrez Gene ID: | 69206 |
Gene Name: | RIKEN cDNA 2010016I18 gene |
Gene Aliases: |
- |
Location: |
Chr.3:106481986-106485913 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2010016I18Rik | NR_033207.1 | 2 | 971 | |
| AK040034.1 | 1 | 882 | BAE20566.1 | |
| AK079963.1 | ||||
| AK080971.1 | 2 | 972 | BAE43373.1 | |
| AK140363.1 | 2 | 972 | BAE24354.1 |
Target Gene Details
Entrez Gene ID: | 72121 |
Gene Name: | DENN/MADD domain containing 2D |
Gene Aliases: |
2010308M01Rik, AI451421 |
Location: |
Chr.3:106482431-106503030 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd2d | NM_001093754.2 | 1 | 143 | NP_001087223.1 |
| NM_001278941.1 | 1 | 124 | NP_001265870.1 | |
| AK050033.1 | 1 | 125 | ||
| AK157279.1 | 1 | 144 | BAE34027.1 |

Genomic Map