Assay Details
Target Gene Details
Entrez Gene ID: | 18441 |
Gene Name: | purinergic receptor P2Y, G-protein coupled 1 |
Gene Aliases: |
P2Y1 |
Location: |
Chr.3:61002785-61008982 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2ry1 | NM_001282016.1 | 1 | 1509 | NP_001268945.1 |
| NM_008772.5 | 1 | 1509 | NP_032798.1 | |
| AK030759.1 | 1 | 1499 | BAC27125.1 | |
| AK033660.1 | 1 | 1495 | BAC28413.1 | |
| AK036611.1 | 1 | 1500 | BAC29506.1 | |
| BC138634.1 | 1 | 1370 | ||
| BC138635.1 | 1 | 1370 | ||
| U22829.1 | 1 | 883 | AAA91302.1 |
More Information
Set Membership: |
|

Genomic Map