Assay Details
Target Gene Details
Entrez Gene ID: | 24136 |
Gene Name: | zinc finger E-box binding homeobox 2 |
Gene Aliases: |
9130203F04Rik, D130016B08Rik, SIP1, Zfhx1b, Zfx1b, Zfxh1b |
Location: |
Chr.2:44983512-45113279 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zeb2 | NM_015753.4 | 1 | 157 | NP_056568.2 |
| AJ535779.1 | ||||
| AJ535780.1 | 1 | 123 | ||
| AJ535782.1 | 1 | 87 | ||
| AK031541.1 | 1 | 103 | BAC27441.1 | |
| AK135557.1 | BAE22581.1 | |||
| CB245498.1 | 1 | 157 |
Target Gene Details
Entrez Gene ID: | 433424 |
Gene Name: | zinc finger E-box binding homeobox 2, opposite strand |
Gene Aliases: |
Gm13476, OTTMUSG00000012358 |
Location: |
Chr.2:45111138-45114087 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zeb2os | NR_110571.1 | |||
| NR_110572.1 |
More Information
Set Membership: |
|

Genomic Map