Assay Details
Target Gene Details
Entrez Gene ID: | 24136 |
Gene Name: | zinc finger E-box binding homeobox 2 |
Gene Aliases: |
9130203F04Rik, D130016B08Rik, SIP1, Zfhx1b, Zfx1b, Zfxh1b |
Location: |
Chr.2:44983512-45113279 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zeb2 | NM_001289521.1 | 9 | 6943 | NP_001276450.1 |
| NM_015753.4 | 10 | 7280 | NP_056568.2 | |
| AK033626.1 | 1 | 707 | ||
| AK042553.1 | 1 | 423 | ||
| AK043069.1 | 1 | 814 | ||
| AK044197.1 | 1 | 552 | ||
| AK046644.1 | 1 | 513 | ||
| AK052619.1 | 1 | 294 | ||
| AK141542.1 | 1 | 2575 | BAE24726.1 |
More Information
Set Membership: |
|

Genomic Map