Assay Details
Target Gene Details
Entrez Gene ID: | 12428 |
Gene Name: | cyclin A2 |
Gene Aliases: |
AA408589, Ccn-1, Ccn1, Ccna, CycA2, Cyca |
Location: |
Chr.3:36564865-36571996 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccna2 | NM_009828.2 | 8 | 2575 | NP_033958.2 |
| AK044924.1 | 8 | 2578 | BAC32144.1 | |
| BC052730.1 | 8 | 2523 | AAH52730.1 | |
| Z26580.1 | 8 | 2532 | CAA81331.1 |
Target Gene Details
Entrez Gene ID: | 50911 |
Gene Name: | exosome component 9 |
Gene Aliases: |
PM/Scl-75, Pmscl1, RRP45, p5, p6 |
Location: |
Chr.3:36552606-36565727 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exosc9 | NM_019393.2 | NP_062266.1 | ||
| AF152841.1 | AAF73218.1 | |||
| AF152842.1 | AAF73219.1 | |||
| AK010253.1 | ||||
| AK011636.1 | BAB27749.1 | |||
| AK076096.1 | ||||
| BC005622.1 | AAH05622.1 | |||
| BC052156.1 | AAH52156.1 | |||
| BC158075.1 |
More Information
Set Membership: |
|

Genomic Map