Assay Details
Target Gene Details
Entrez Gene ID: | 70604 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 14 |
Gene Aliases: |
5730496F10Rik |
Location: |
Chr.3:137867675-137916563 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb14 | NM_001033155.1 | 1 | 259 | NP_001028327.1 |
| AK017735.1 | 1 | 122 | ||
| AK160961.1 | 1 | 48 | BAE36117.1 | |
| BC117717.1 | 1 | 60 | ||
| BC117718.1 | 1 | 60 |

Genomic Map