Assay Details
Target Gene Details
Entrez Gene ID: | 14865 |
Gene Name: | glutathione S-transferase, mu 4 |
Gene Aliases: |
1110004G14Rik, GSTM7-7, Gstb-4, Gstb4 |
Location: |
Chr.3:108040408-108045080 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gstm4 | NM_001160411.1 | 7 | 1307 | NP_001153883.1 |
| NM_026764.3 | 8 | 1512 | NP_081040.1 | |
| AF464943.1 | 8 | 1342 | AAL76248.1 | |
| AF501320.1 | 8 | 1328 | AAM67419.1 | |
| AI842977.1 | 1 | 173 | ||
| AK003418.1 | 7 | 1308 | ||
| BC030444.1 | 8 | 1459 | AAH30444.1 |
More Information
Set Membership: |
|

Genomic Map