Assay Details
Target Gene Details
Entrez Gene ID: | 67437 |
Gene Name: | signal sequence receptor, gamma |
Gene Aliases: |
0610038P07Rik, AL022999, AU022074, AW553833, TRAPG |
Location: |
Chr.3:65379657-65392553 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssr3 | NM_026155.3 | 5 | 1303 | NP_080431.1 |
| AK002818.1 | 5 | 1304 | BAB22381.1 | |
| AK012912.1 | 1 | 575 | ||
| AK076235.1 | 5 | 1336 | BAC36268.1 | |
| AK088555.1 | 5 | 1308 | BAC40420.1 | |
| AK150255.1 | 5 | 1297 | BAE29416.1 | |
| AK162235.1 | 3 | 1007 | BAE36806.1 | |
| BC011111.1 | 5 | 1310 | AAH11111.1 |

Genomic Map