Assay Details
Target Gene Details
Entrez Gene ID: | 20257 |
Gene Name: | stathmin-like 2 |
Gene Aliases: |
AI159727, SCG10, Scgn10, Stmb2 |
Location: |
Chr.3:8509527-8561604 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stmn2 | NM_025285.2 | 5 | 1007 | NP_079561.1 |
| AK003659.1 | 5 | 1007 | BAB22921.1 | |
| AK013020.1 | 5 | 1008 | BAB28602.1 | |
| AK149212.1 | 5 | 1032 | BAE28766.1 | |
| BC026538.1 | 5 | 969 | AAH26538.1 |

Genomic Map