Assay Details
Target Gene Details
Entrez Gene ID: | 24136 |
Gene Name: | zinc finger E-box binding homeobox 2 |
Gene Aliases: |
9130203F04Rik, D130016B08Rik, SIP1, Zfhx1b, Zfx1b, Zfxh1b |
Location: |
Chr.2:44983512-45113279 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zeb2 | NM_001289521.1 | 2 | 396 | NP_001276450.1 |
| NM_015753.4 | 3 | 733 | NP_056568.2 | |
| AF033116.1 | 2 | 248 | AAD56590.1 | |
| AK012377.1 | 2 | 204 | ||
| AK031541.1 | 4 | 830 | BAC27441.1 | |
| AK122312.1 | 3 | 306 | BAC65594.1 | |
| AK135557.1 | 3 | 543 | BAE22581.1 | |
| AK141157.1 | 2 | 364 | BAE24572.1 | |
| AK157561.1 | 3 | 456 | ||
| BC060699.1 | 3 | 470 | AAH60699.1 | |
| CB245498.1 | ||||
| CJ130482.1 |
More Information
Set Membership: |
|

Genomic Map