Assay Details
Target Gene Details
Entrez Gene ID: | 99480 |
Gene Name: | deoxynucleotidyltransferase, terminal, interacting protein 2 |
Gene Aliases: |
4930588M11Rik, AA408582, AU014960, HSU15552 |
Location: |
Chr.3:122274414-122285271 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnttip2 | NM_153806.1 | 2 | 1132 | NP_722501.1 |
| AK016368.1 | 2 | 1125 | ||
| AK133904.1 | 2 | 1151 | ||
| AK168536.1 | 2 | 1151 | BAE40414.1 | |
| AK214673.1 | ||||
| BC028305.1 | 2 | 1132 | AAH28305.1 |

Genomic Map