Assay Details
Target Gene Details
Entrez Gene ID: | 68563 |
Gene Name: | dolichyl-phosphate mannosyltransferase polypeptide 3 |
Gene Aliases: |
1110001H19Rik, AV117483 |
Location: |
Chr.3:89259263-89267079 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpm3 | NM_026767.4 | 2 | 323 | NP_081043.1 |
| AI527477.1 | 1 | 267 | ||
| AK003223.1 | 2 | 233 | BAB22652.1 | |
| CJ237809.1 | 2 | 323 |
More Information
Set Membership: |
|

Genomic Map