Assay Details
Target Gene Details
Entrez Gene ID: | 66460 |
Gene Name: | SYS1 Golgi-localized integral membrane protein homolog (S. cerevisiae) |
Gene Aliases: |
2610042O14Rik, AI266964, C87389 |
Location: |
Chr.2:164456981-164479637 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sys1 | NM_025575.3 | 4 | 835 | NP_079851.2 |
| AK003071.1 | 4 | 828 | BAB22547.1 | |
| AK005249.1 | 4 | 738 | ||
| AK005510.1 | 4 | 772 | BAB24089.1 | |
| AK007882.1 | 4 | 834 | ||
| AK009684.1 | 4 | 799 | ||
| AK011753.1 | 4 | 831 | ||
| AK013220.1 | 4 | 808 | BAB28722.1 | |
| AK017524.1 | 3 | 667 | ||
| AK032073.1 | ||||
| AK032098.1 | 3 | 670 | ||
| AK075770.1 | 4 | 800 | BAC35944.1 | |
| AK148305.1 | 4 | 773 | BAE28468.1 | |
| AK210170.1 | 1 | 189 | ||
| AK216375.1 | 1 | 193 | ||
| AK219257.1 | 1 | 193 | ||
| BC061084.1 | 3 | 709 | AAH61084.1 |
More Information
Set Membership: |
|

Genomic Map