Assay Details
Target Gene Details
Entrez Gene ID: | 11522 |
Gene Name: | alcohol dehydrogenase 1 (class I) |
Gene Aliases: |
ADH-AA, AI194826, Adh-1, Adh-1-t, Adh-1e, Adh-1t, Adh-3e, Adh1-e, Adh1-t, Adh1tl, Adh3-e |
Location: |
Chr.3:138277645-138290691 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adh1 | NM_007409.2 | 6 | 851 | NP_031435.1 |
| AK146094.1 | 6 | 852 | BAE26897.1 | |
| BC013477.1 | 6 | 851 | AAH13477.1 | |
| BC054467.1 | 6 | 831 | AAH54467.1 | |
| M22611.1 | 1 | 110 | AAA37181.1 |
More Information
Set Membership: |
|

Genomic Map