Assay Details
Target Gene Details
Entrez Gene ID: | 14700 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 10 |
Gene Aliases: |
AV006524 |
Location: |
Chr.4:59035156-59041899 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng10 | NM_025277.3 | 3 | 830 | NP_079553.1 |
| AK003187.1 | 3 | 768 | ||
| BB434753.1 | ||||
| BC023670.1 | 3 | 773 | AAH23670.1 | |
| BC055434.1 | 4 | 755 | AAH55434.1 |
More Information
Set Membership: |
|

Genomic Map